Anti-C3/C3b Mouse Monoclonal Antibody [clone: 2B5]
Catalog # ABCA_AB315208-100U
Supplier: Abcam
New Product
Specifications
- Antibody type:Primary
- Antigen name:CPAMD1
- Antigen symbol:C3/C3b
- Clonality:Monoclonal
- Clone:2B5
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen synonyms:C3|Complement C3|C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
- Molecular weight:187148 Da
- Storage temperature:Store at 4 °C short term (1 - 2 weeks). Store at −20 °C. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping temperature:4 °C
- Immunogen:Recombinant Full Length Protein corresponding to Human C3
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [2B5] to C3/C3b.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: C3/C3b
Clonality: Monoclonal
Clone: 2B5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human