Anti-Proteasome 20S alpha 1+2+3+5+6+7 Mouse Monoclonal Antibody [clone: MCP231]
Catalog # ABCA_AB22674-50UL
Supplier: Abcam
New Product
Specifications
- Antibody type:Primary
- Antigen name:HC2
- Antigen symbol:Proteasome 20S alpha 1+2+3+5+6+7
- Clonality:Monoclonal
- Clone:MCP231
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Size:50 µl
- Epitope:Reacts with the peptide sequence TVWSPQGRLHQVEYAMEA encompassing the prosbox I motif common to alpha type subunits, although not necessarily identical in all. In general this motif is phylogenetically preserved.
- Form:Liquid
- Antigen synonyms:NU|PROS30|PSC2|PSMA1|Proteasome subunit alpha type-1|30 kDa prosomal protein|Macropain subunit C2|Multicatalytic endopeptidase complex subunit C2|Proteasome component C2|Proteasome nu chain|Proteasome subunit alpha-6|PROS-30|alpha-6
- Storage buffer:PBS
- Molecular weight:29556 Da
- Storage temperature:Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped at +4 °C
- Immunogen:Full Length Protein corresponding to Human PSMA1. The exact immunogen used to generate this antibody is proprietary information.
- Tested applications:ICC/IF
- Purification:Protein G Affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [MCP231] to Proteasome 20S alpha 1+2+3+5+6+7.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Proteasome 20S alpha 1+2+3+5+6+7
Clonality: Monoclonal
Clone: MCP231
Conjugation:
Epitope: Reacts with the peptide sequence TVWSPQGRLHQVEYAMEA encompassing the prosbox I motif common to alpha type subunits, although not necessarily identical in all. In general this motif is phylogenetically preserved.
Host: Mouse
Isotype: IgG1
Reactivity: Human