Anti-KAP1 (phospho S824) Rabbit Monoclonal Antibody [clone: EPR5248]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR5248] to KAP1 (phospho S824) - BSA and Azide free.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: KAP1
Clonality: Monoclonal
Clone: EPR5248
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- ABCA_AB215549-100U
- ABCA_AB215549-1MG
- Antigen symbol:
- KAP1
- KAP1
- Antigen name:
- RNF96
- RNF96
- Antigen synonyms:
- TIF1B|TRIM28|Transcription intermediary factor 1-beta|TIF1-beta|E3 SUMO-protein ligase TRIM28|KRAB-associated protein 1|KRAB-interacting protein 1|Nuclear corepressor KAP-1|RING finger protein 96|RING-type E3 ubiquitin transferase TIF1-beta|Tripartite motif-containing protein 28|KAP-1|KRIP-1
- TIF1B|TRIM28|Transcription intermediary factor 1-beta|TIF1-beta|E3 SUMO-protein ligase TRIM28|KRAB-associated protein 1|KRAB-interacting protein 1|Nuclear corepressor KAP-1|RING finger protein 96|RING-type E3 ubiquitin transferase TIF1-beta|Tripartite motif-containing protein 28|KAP-1|KRIP-1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR5248
- EPR5248
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Modification:
- phospho S824
- phospho S824
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular weight:
- 88550 Da
- 88550 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage temperature:
- Store at 4 °C. Do not freeze.
- Store at 4 °C. Do not freeze.
- Shipping temperature:
- 4 °C
- 4 °C
- Tested applications:
- Dot
- Dot
Specifications
Frequently Bought Together