Anti-Neuropeptide Y Mouse Monoclonal Antibody [clone: 8]
ABCA_AB112473-100U
: Abcam
New Product
- Antibody type:Primary
- Antigen name:Pro-neuropeptide Y
- Antigen symbol:Neuropeptide Y
- Clonality:Monoclonal
- Clone:8
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Mouse,Human
- Size:100 µg
- Epitope:The epitope of ab112473 is probably around the mid-molecular region of Neuropeptide Y
- Form:Liquid
- Antigen synonyms:NPY
- Molecular weight:10851 Da
- Storage temperature:Store at 4 °C short term (1 - 2 weeks). Store at −20 or −80 °C. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping temperature:4 °C
- Immunogen:Synthetic Peptide within Human NPY. The exact immunogen used to generate this antibody is proprietary information.
- Tested applications:IHC-P
- Purification:Protein G affinity purification
- Packaging:Vial
Mouse monoclonal [8] to Neuropeptide Y.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Neuropeptide Y
Clonality: Monoclonal
Clone: 8
Conjugation:
Epitope: The epitope of ab112473 is probably around the mid-molecular region of Neuropeptide Y
Host: Mouse
Isotype: IgG1
Reactivity: Mouse;Human