Anti-Siglec 7 Mouse Monoclonal Antibody [clone: 6-434] (PE (Phycoerythrin))
Catalog # ABCA_AB200558-100T
Supplier: Abcam
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD328
- Antigen symbol:Siglec 7
- Clonality:Monoclonal
- Clone:6-434
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Size:100 Tests
- Form:Liquid
- Antigen synonyms:AIRM1|SIGLEC7|Sialic acid-binding Ig-like lectin 7|Siglec-7|Adhesion inhibitory receptor molecule 1|CDw328|D-siglec|QA79 membrane protein|p75|AIRM-1
- Molecular weight:51143 Da
- Storage temperature:Store at 4 °C. Do not freeze. Store in the dark.
- Shipping temperature:4 °C
- Immunogen:Cell preparation containing SIGLEC7 protein. The exact immunogen used to generate this antibody is proprietary information.
- Purification:Size-exclusion chromatography
- Packaging:Vial
Specifications
About this item
PE Mouse monoclonal [6-434] to Siglec 7, prediluted.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Siglec 7
Clonality: Monoclonal
Clone: 6-434
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human