Anti-CCN3 Rabbit Polyclonal Antibody
Catalog # ABCA_AB137677-100U
Supplier: Abcam
New Product
Specifications
- Antibody type:Primary
- Antigen name:IGFBP9
- Antigen symbol:CCN3
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen synonyms:NOV|NOVH|CCN family member 3|Cellular communication network factor 3|Insulin-like growth factor-binding protein 9|Nephro blastoma-overexpressed gene protein homolog|Protein NOV homolog|IBP-9|IGF-binding protein 9|IGFBP-9
- Molecular weight:39162 Da
- Storage temperature:Upon delivery aliquot. Store at –20 or –80 °C. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping temperature:Shipped at +4 °C
- Immunogen:Recombinant Fragment Protein within Human CCN3 aa 50 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
- Tested applications:ICC/IF, IHC-P
- Purification:Immunogen Affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to CCN3.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CCN3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse