Anti-KIR3DL1 Mouse Monoclonal Antibody [clone: 6D9F6]
Catalog # ABCA_AB233740-100U
Supplier: Abcam
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD158e
- Antigen symbol:KIR3DL1
- Clonality:Monoclonal
- Clone:6D9F6
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:NKAT3|NKB1|Killer cell immunoglobulin-like receptor 3DL1|CD158 antigen-like family member E|HLA-BW4-specific inhibitory NK cell receptor|Natural killer-associated transcript 3|p70 natural killer cell receptor clones CL-2/CL-11|NKAT-3|p70 NK receptor CL-2/CL-11
- Molecular weight:49098 Da
- Storage temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping temperature:4 °C
- Immunogen:Recombinant Fragment Protein within Human KIR3DL1 aa 1-350. The exact immunogen used to generate this antibody is proprietary information.
- Tested applications:IHC-P
- Purification:Protein G affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [6D9F6] to KIR3DL1.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: KIR3DL1
Clonality: Monoclonal
Clone: 6D9F6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat;Human;African green monkey