Anti-DDX39 Rabbit Monoclonal Antibody [clone: EPR13507(B)] (PE)
Catalog # ABCA_AB225253-100U
Supplier: Abcam
New Product
Specifications
- Antibody type:Primary
- Antigen name:DDX39A
- Antigen symbol:DDX39
- Clonality:Monoclonal
- Clone:EPR13507(B)
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Form:Liquid
- Antigen synonyms:ATP-dependent RNA helicase DEAD box protein 39|Nuclear RNA helicase URH49
- Molecular weight:49130 Da
- Storage temperature:Shipped at +4 °C. Upon delivery aliquot. Store at +4 °C. Do Not Freeze. Store In the Dark.
- Concentration:0.5 mg/ml
- Shipping temperature:+4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information.
- Tested applications:Flow Cyt (Intra)
- Purification:Protein A Affinity purification
- Packaging:Vial
Specifications
About this item
PE Rabbit monoclonal [EPR13507(B)] to DDX39.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: DDX39
Clonality: Monoclonal
Clone: EPR13507(B)
Conjugation: PE
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human