Anti-FGFR3 (phospho Y724) Rabbit Monoclonal Antibody [clone: EPR2281(3)] (Alexa Fluor® 488)
Catalog # ABCA_AB237164-100U
Supplier: Abcam
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD333
- Antigen symbol:FGFR3
- Clonality:Monoclonal
- Clone:EPR2281(3)
- Conjugation:Alexa Fluor® 488
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Size:100 µl
- Form:Liquid
- Antigen synonyms:JTK4|Fibroblast growth factor receptor 3|FGFR-3
- Modification:phospho Y724
- Molecular weight:87710 Da
- Storage temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle. Stable for 12 months at −20 °C. Store in the dark.
- Concentration:0.5 mg/ml
- Shipping temperature:4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information
- Tested applications:Flow Cyt (Intra), ICC/IF
- Purification:Protein A affinity purification
- Packaging:Vial
Specifications
About this item
Alexa Fluor® 488 Rabbit monoclonal [EPR2281(3)] to FGFR3 (phospho Y724).
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: FGFR3
Clonality: Monoclonal
Clone: EPR2281(3)
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human