Order Entry
Canada
ContactUsLinkComponent
Anti-cGMP Sheep Polyclonal Antibody
Anti-cGMP Sheep Polyclonal Antibody
Catalog # ABCA_AB836-100UL
Supplier:  Abcam
New Product
Anti-cGMP Sheep Polyclonal Antibody
Catalog # ABCA_AB836-100UL
Supplier:  Abcam
Supplier Number:  AB836-100UL
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

About this item

Sheep polyclonal to cGMP

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cytokeratin
Clonality: Monoclonal
Clone: MNF116
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human