Anti-Carbonic Anhydrase 12/CA12 Rabbit Polyclonal Antibody
ABCA_AB244309-100U
: Abcam
New Product
- Antibody type:Primary
- Antigen name:Carbonic anhydrase 12
- Antigen symbol:Carbonic Anhydrase 12/CA12
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Size:100 µl
- Form:Liquid
- Antigen synonyms:Carbonate dehydratase XII|Carbonic anhydrase XII|Tumor antigen HOM-RCC-3.1.3|CA-XII|CA12
- Molecular weight:39451 Da
- Storage temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Shipping temperature:4 °C
- Immunogen:Recombinant Fragment Protein within Human CA12 aa 150-300. The exact immunogen used to generate this antibody is proprietary information.
- Tested applications:ICC/IF, IHC-P
- Purification:Immunogen affinity purification
- Packaging:Vial
Rabbit polyclonal to Carbonic Anhydrase 12/CA12.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Carbonic Anhydrase 12/CA12
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human