Anti-Lysozyme Rabbit Monoclonal Antibody [clone: SP329]
ABCA_AB242420-100U
: Abcam
New Product
- Antibody type:Primary
- Antigen name:LZM
- Antigen symbol:Lysozyme
- Clonality:Monoclonal
- Clone:SP329
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:LYZ|Lysozyme C
- Molecular weight:16537 Da
- Storage temperature:Store at 4 °C. Do not freeze.
- Shipping temperature:4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information
- Tested applications:Flow Cyt (Intra), IHC-P
- Purification:Protein A affinity purification
- Packaging:Vial
Rabbit monoclonal [SP329] to Lysozyme - BSA and Azide free.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Lysozyme
Clonality: Monoclonal
Clone: SP329
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human