Anti-alpha 1 Sodium Potassium ATPase Mouse Monoclonal Antibody [clone: M8-P1-A3]
ABCA_AB2872-100UG
: Abcam
New Product
- Size:100 µg
- Packaging:Vial
Mouse monoclonal [M8-P1-A3] to alpha 1 Sodium Potassium ATPase.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: alpha 1 Sodium Potassium ATPase
Clonality: Monoclonal
Clone: M8-P1-A3
Conjugation:
Epitope: This antibody recognizes an epitope between amino acid residues 496 - 506 of lamb kidney sodium/potassium ATPase
Host: Mouse
Isotype: IgG1
Reactivity: Human