Anti-eRF3/GSPT1 Rabbit Polyclonal Antibody
Catalogue # ABCA_AB126090-50UL
Fournisseur: Abcam
New Product
Spécifications
- Antibody type:Primary
- Antigen name:ERF3A
- Antigen symbol:eRF3/GSPT1
- Clonality:Polyclonal
- Clone:0
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Antigen synonyms:GSPT1|Eukaryotic peptide chain release factor GTP-binding subunit Eukaryotic peptide chain release factor subunit 3a|G1 to S phase transition protein 1 homolog
- Molecular weight:55756 Da
- Storage temperature:Upon delivery aliquot. Store at –20 or –80 °C. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping temperature:Shipped at +4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information.
- Tested applications:IHC-P
- Purification:Immunogen Affinity purification
- Packaging:Vial
Spécifications
À propos de cet article
Rabbit polyclonal to eRF3/GSPT1.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: eRF3/GSPT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human