ContactUsLinkComponent
Anti-E tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-E tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # ABCA_AB3415-250UG
Supplier:  Abcam
New Product
Anti-E tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # ABCA_AB3415-250UG
Supplier:  Abcam
Supplier Number:  AB3415-250UG
New Product
Specifications
  • Size:
    250 µg
  • Packaging:
    Vial
Specifications
About this item
HRP Rabbit polyclonal to E tag.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary Antigen: E tag Clonality: Polyclonal Clone: Conjugation: HRP (Horseradish Peroxidase) Epitope: Host: Rabbit Isotype: IgG Reactivity: