Anti-Integrin beta 1 Mouse Monoclonal Antibody [clone: M032]
ABCA_AB253282-100U
: Abcam
New Product
- Antibody type:Primary
- Antigen name:CD29
- Antigen symbol:Integrin beta 1
- Clonality:Monoclonal
- Clone:M032
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen synonyms:FNRB|MDF2|MSK12|ITGB1|Integrin beta-1|Fibronectin receptor subunit beta|Glycoprotein IIa|VLA-4 subunit beta|GPIIA
- Molecular weight:88415 Da
- Storage temperature:Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
- Shipping temperature:+4 °C
- Immunogen:Recombinant Fragment Protein within Human ITGB1. The exact immunogen used to generate this antibody is proprietary information.
- Tested applications:ICC/IF
- Purification:Protein G Affinity purification
- Packaging:Vial
Mouse monoclonal [M032] to Integrin beta 1 - Extracellular domain.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Integrin beta 1
Clonality: Monoclonal
Clone: M032
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human