Anti-Carbonic Anhydrase 9/CA9 Rabbit Monoclonal Antibody [clone: SP106]
: Abcam
New Product
Rabbit monoclonal [SP106] to Carbonic Anhydrase 9/CA9 - BSA and Azide free.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Carbonic Anhydrase 9/CA9
Clonality: Monoclonal
Clone: SP106
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- ABCA_AB243910-100U
- ABCA_AB243910-1MG
- Antigen symbol:
- Carbonic Anhydrase 9/CA9
- Carbonic Anhydrase 9/CA9
- Antigen name:
- G250
- G250
- Antigen synonyms:
- MN|CA9|Carbonic anhydrase 9|Carbonate dehydratase IX|Carbonic anhydrase IX|Membrane antigen MN|P54/58N|Renal cell carcinoma-associated antigen pMW1|CA-IX|CAIX|RCC-associated antigen G250
- MN|CA9|Carbonic anhydrase 9|Carbonate dehydratase IX|Carbonic anhydrase IX|Membrane antigen MN|P54/58N|Renal cell carcinoma-associated antigen pMW1|CA-IX|CAIX|RCC-associated antigen G250
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- SP106
- SP106
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ImmunoChemistry:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular weight:
- 49698 Da
- 49698 Da
- Purification:
- Protein A/G affinity purification
- Protein A/G affinity purification
- Storage temperature:
- Store at 4 °C. Do not freeze.
- Store at 4 °C. Do not freeze.
- Shipping temperature:
- 4 °C
- 4 °C
- Tested applications:
- IHC-P
- IHC-P
Frequently Bought Together