Anti-NFkB Inducing Kinase NIK Rabbit Monoclonal Antibody [clone: 3H31L48]
Catalog # ABCA_AB203568-100U
Supplier: Abcam
New Product
Specifications
- Antibody type:Primary
- Antigen name:NIK
- Antigen symbol:NFkB Inducing Kinase NIK
- Clonality:Monoclonal
- Clone:3H31L48
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:MAP3K14|Mitogen-activated protein kinase kinase kinase 14|NF-kappa-beta-inducing kinase|Serine/threonine-protein kinase Hs
- Molecular weight:104042 Da
- Storage temperature:Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
- Concentration:0.5 mg/ml
- Shipping temperature:+4 °C
- Immunogen:Recombinant Fragment Protein within Human MAP3K14 aa 700-900. The exact immunogen used to generate this antibody is proprietary information.
- Tested applications:ICC/IF
- Purification:Protein A Affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit monoclonal [3H31L48] to NFkB Inducing Kinase NIK.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: NFkB Inducing Kinase NIK
Clonality: Monoclonal
Clone: 3H31L48
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human