Anti-GABA Transporter 1/GAT 1 Rabbit Monoclonal Antibody [clone: EPR24202-20]
: Abcam
New Product
Rabbit monoclonal [EPR24202-20] to GABA Transporter 1 / GAT 1 - BSA and Azide free.
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: GABA Transporter 1 / GAT 1
Clonality: Monoclonal
Clone: EPR24202-20
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
- Catalog No:
- ABCA_AB277642-1MG
- ABCA_AB277642-100U
- Antigen symbol:
- GABA Transporter 1 / GAT 1
- GABA Transporter 1 / GAT 1
- Antigen name:
- Gabt1
- Gabt1
- Antigen synonyms:
- Gat-1|Gat1|Slc6a1|Sodium- and chloride-dependent GABA transporter 1|Solute carrier family 6 member 1
- Gat-1|Gat1|Slc6a1|Sodium- and chloride-dependent GABA transporter 1|Solute carrier family 6 member 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR24202-20
- EPR24202-20
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular weight:
- 67001 Da
- 67001 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage temperature:
- Store at +4 °C.
- Store at +4 °C.
- Shipping temperature:
- Shipped at +4 °C
- Shipped at +4 °C
- Tested applications:
- ICC/IF, IHC-Fr, IHC-P
- ICC/IF, IHC-Fr, IHC-P
Frequently Bought Together