Anti-CC2D1A Rabbit Monoclonal Antibody [clone: EPR18421]
: Abcam
New Product
Rabbit monoclonal [EPR18421] to CC2D1A.
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CC2D1A
Clonality: Monoclonal
Clone: EPR18421
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
- Catalog No:
- ABCA_AB191472-1ML
- ABCA_AB191472-100U
- Antigen symbol:
- CC2D1A
- CC2D1A
- Antigen name:
- AKI1
- AKI1
- Antigen synonyms:
- LGD2|Coiled-coil and C2 domain-containing protein 1A|Akt kinase-interacting protein 1|Five prime repressor element under dual repression-binding protein 1|Putative NF-kappa-B-activating protein 023N|FRE under dual repression-binding protein 1|Freud-1
- LGD2|Coiled-coil and C2 domain-containing protein 1A|Akt kinase-interacting protein 1|Five prime repressor element under dual repression-binding protein 1|Putative NF-kappa-B-activating protein 023N|FRE under dual repression-binding protein 1|Freud-1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR18421
- EPR18421
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular weight:
- 104062 Da
- 104062 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage temperature:
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze/thaw cycle.
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze/thaw cycle.
- Shipping temperature:
- Shipped at +4 °C
- Shipped at +4 °C
- Tested applications:
- Flow Cyt (Intra), ICC/IF
- Flow Cyt (Intra), ICC/IF
Frequently Bought Together