Anti-MCAF1 Rabbit Polyclonal Antibody
Catalog # ABCA_AB84497-100UL
Supplier: Abcam
New Product
Specifications
- Antibody type:Primary
- Antigen name:MCAF
- Antigen symbol:MCAF1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Size:100 µl
- Form:Liquid
- Antigen synonyms:ATF7IP|Activating transcription factor 7-interacting protein 1|ATF-interacting protein|ATF7-interacting protein|ATFa-associated modulator|MBD1-containing chromatin-associated factor 1|P621|ATF-IP|hAM
- Storage buffer:BSA, tris buffered saline
- Molecular weight:136394 Da
- Storage temperature:Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:4 °C
- Immunogen:Synthetic Peptide within Human ATF7IP aa 1-100. The exact immunogen used to generate this antibody is proprietary information.
- Tested applications:IHC-P
- Purification:Immunogen affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to MCAF1.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: MCAF1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human