Anti-LHX5 Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Rabbit Polyclonal LHX5 antibody. Suitable for IHC-P and reacts with Human samples. Cited in 1 publication. Immunogen corresponding to Recombinant Fragment Protein within Human LHX5 aa 100-200.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: LHX5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Size:100 µl
- Packaging:Vial
- Clonality:Polyclonal
- Purification:Immunogen affinity purification
- Antibody type:Primary
- Form:Liquid
- Isotype:IgG
- Storage temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
- Host:Rabbit
- Molecular weight:44406 Da
- Shipping temperature:4 °C
- Antigen name:LIM/homeobox protein Lhx5
- Cat. No.:ABCA_AB187975-100U
- Reactivity:Human
- Antigen synonyms:LIM homeobox protein 5
- Immunogen:Recombinant Fragment Protein within Human LHX5 aa 100-200. The exact immunogen used to generate this antibody is proprietary information.
- ImmunoChemistry:Yes
- Tested applications:IHC-P
- Antigen symbol:LHX5
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



