Order Entry
Canada
ContactUsLinkComponent
Anti-Ubiquitin (Linkage-specific K48) Rabbit Monoclonal Antibody [clone: EP8589] (Alexa Fluor® 568)
Catalogue # ABCA_AB312542-100U
Fournisseur:  Abcam
New Product
Anti-Ubiquitin (Linkage-specific K48) Rabbit Monoclonal Antibody [clone: EP8589] (Alexa Fluor® 568)
Catalogue # ABCA_AB312542-100U
Fournisseur:  Abcam
Supplier Number:  AB312542-100UL
New Product

À propos de cet article

Alexa Fluor® 568 Rabbit monoclonal [EP8589] to Ubiquitin (linkage-specific K48).

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Ubiquitin
Clonality: Monoclonal
Clone: EP8589
Conjugation: Alexa Fluor® 568
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: