Anti-cleaved N-terminal DFNA5/GSDME Rabbit Monoclonal Antibody [clone: EPR20867-248]
: Abcam
New Product
Rabbit monoclonal [EPR20867-248] to cleaved N-terminal DFNA5 / GSDME - BSA and Azide free.
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: cleaved N-terminal DFNA5 / GSDME
Clonality: Monoclonal
Clone: EPR20867-248
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- ABCA_AB256165-100U
- ABCA_AB256165-1MG
- Antigen symbol:
- cleaved N-terminal DFNA5 / GSDME
- cleaved N-terminal DFNA5 / GSDME
- Antigen name:
- DFNA5
- DFNA5
- Antigen synonyms:
- ICERE1|GSDME|Gasdermin-E|Inversely correlated with estrogen receptor expression 1|Non-syndromic hearing impairment protein 5|ICERE-1
- ICERE1|GSDME|Gasdermin-E|Inversely correlated with estrogen receptor expression 1|Non-syndromic hearing impairment protein 5|ICERE-1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR20867-248
- EPR20867-248
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage temperature:
- Store at +4 °C. Do not freeze.
- Store at +4 °C. Do not freeze.
- Shipping temperature:
- Shipped at +4 °C
- Shipped at +4 °C
Frequently Bought Together