Order Entry
Canada
ContactUsLinkComponent
Anti-ATP5G1/G2/G3 Rabbit Monoclonal Antibody [clone: EPR13908] (PE (Phycoerythrin))
Anti-ATP5G1/G2/G3 Rabbit Monoclonal Antibody [clone: EPR13908] (PE (Phycoerythrin))
Catalog # ABCA_AB319363-100U
Supplier:  Abcam
New Product
Anti-ATP5G1/G2/G3 Rabbit Monoclonal Antibody [clone: EPR13908] (PE (Phycoerythrin))
Catalog # ABCA_AB319363-100U
Supplier:  Abcam
Supplier Number:  AB319363-100UL
New Product

Specifications

  • Size:
    100 µl
  • Packaging:
    Vial

Specifications

About this item

PE Rabbit monoclonal [EPR13908] to ATP5G1/G2/G3.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ATP5G1/G2/G3
Clonality: Monoclonal
Clone: EPR13908
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: