Anti-Cripto1/CRIPTO Rabbit Monoclonal Antibody [clone: EPR22230-72]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR22230-72] to Cripto1/CRIPTO - BSA and Azide free (Capture).
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Cripto1/CRIPTO
Clonality: Monoclonal
Clone: EPR22230-72
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB259567-1MG
- ABCA_AB259567-100U
- Antigen symbol:
- Cripto1/CRIPTO
- Cripto1/CRIPTO
- Antigen name:
- CRIPTO-1
- CRIPTO-1
- Antigen synonyms:
- TDGF1|CRIPTO|Protein Cripto|Cripto-1 growth factor|Epidermal growth factor-like cripto protein CR1|Teratocarcinoma-derived growth factor 1|CRGF
- TDGF1|CRIPTO|Protein Cripto|Cripto-1 growth factor|Epidermal growth factor-like cripto protein CR1|Teratocarcinoma-derived growth factor 1|CRGF
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR22230-72
- EPR22230-72
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular weight:
- 21169 Da
- 21169 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage temperature:
- Store at 4 °C
- Store at 4 °C
- Shipping temperature:
- 4 °C
- 4 °C
- Tested applications:
- sELISA
- sELISA
Specifications
Frequently Bought Together