Anti-TRPC1 Rabbit Monoclonal Antibody [clone: EPR1417(2)]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR1417(2)] to TRPC1.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: TRPC1
Clonality: Monoclonal
Clone: EPR1417(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- ABCA_AB192031-100U
- ABCA_AB192031-1ML
- Antigen symbol:
- TRPC1
- TRPC1
- Antigen name:
- TRP1
- TRP1
- Antigen synonyms:
- Short transient receptor potential channel 1|Transient receptor protein 1|TRP-1
- Short transient receptor potential channel 1|Transient receptor protein 1|TRP-1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR1417(2)
- EPR1417(2)
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular weight:
- 91212 Da
- 91212 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Concentration:
- 0.832 mg/ml
- 0.832 mg/ml
- Storage temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Shipping temperature:
- 4 °C
- 4 °C
Specifications
Frequently Bought Together