Anti-Cytochrome P450 1A2 Rabbit Monoclonal Antibody [clone: EPR6138(2)]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR6138(2)] to Cytochrome P450 1A2 - BSA and Azide free.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Cytochrome P450 1A2
Clonality: Monoclonal
Clone: EPR6138(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB249045-100U
- ABCA_AB249045-1MG
- Antigen synonyms:
- Cholesterol 25-hydroxylase|Cytochrome P(3)450|Cytochrome P450 4|Cytochrome P450-P3|Hydroperoxy icosatetraenoate dehydratase|CYP1A2
- Cholesterol 25-hydroxylase|Cytochrome P(3)450|Cytochrome P450 4|Cytochrome P450-P3|Hydroperoxy icosatetraenoate dehydratase|CYP1A2
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Host:
- Rabbit
- Rabbit
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular weight:
- 58407 Da
- 58407 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage temperature:
- +4 °C
- +4 °C
- Shipping temperature:
- Shipped at +4 °C. Store at +4 °C. Do Not Freeze.
- Shipped at +4 °C. Store at +4 °C. Do Not Freeze.
- Tested applications:
- Flow Cyt (Intra), ICC/IF
- Flow Cyt (Intra), ICC/IF
Specifications
Frequently Bought Together