Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC Ethynylferrocene (>90%)
TRC Ethynylferrocene (>90%)
TRC Ethynylferrocene (>90%)
Catalog # 77984-519
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Ethynylferrocene (>90%)
Catalog # 77984-519
Supplier:
LGC Standards
Supplier Number:
TRC-E937105-500MG
New Product
CAS Number:
Specifications
CAS No.:
1271-47-2
Description:
Ethynylferrocene (>90%)
Size:
500 mg
Specifications
About this item
Product Details
Product Details