Dr. Ehrenstorfer Perfluorooctanesulfonic acid 13C4 (1,2,3,4-13C4) sodium 50 µg/ml in Methanol:Water
Catalog # 77976-047
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol:Water 96:4
- CAS No.:960315-53-1
- Description:Perfluorooctanesulfonic acid 13C4 (1, 2, 3, 4 - 13C4) sodium 50 µg/ml in Methanol:Water
- Size:1 ml
- Shipping conditions:20 °C
- Formula:¹³C₄C₄F₁₇O₃S·Na
- Formula weight:526,082 g/mol
- Storage temperature:−18 °C
- Certification:ISO 17034