Order Entry
Canada
ContactUsLinkComponent
CHEMets® molybdate Refill 0 to 7 ppm
CHEMets® molybdate Refill 0 to 7 ppm
Catalog # 77674-900
Supplier:  CHEMetrics
New Product
CAS Number:  
CHEMets® molybdate Refill 0 to 7 ppm
Catalog # 77674-900
Supplier:  CHEMetrics
Supplier Number:  R-6702
New Product
CAS Number:  

Specifications

  • Chemical method:
    Catechol
  • Application:
    Industrial water treatment;Power generation;Cooling water systems

Specifications

About this item

This product is a refill for K-6701 molybdate CHEMets visual kit and K-6702 molybdate CHEMets visual kit for the analysis of molybdate in water.It contains 30 self-filling CHEMets ampoules packaged securely in cardboard trays and box. R-6702 molybdate CHEMets refill is compatible with C-6701 molybdate Comparator and C-6702 molybdate Comparator.

  • Method: Catechol