Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 6-(Aminocarbonyl)-3-cyclohexene-1-carboxylic Acid
TRC 6-(Aminocarbonyl)-3-cyclohexene-1-carboxylic Acid
TRC 6-(Aminocarbonyl)-3-cyclohexene-1-carboxylic Acid
Catalog # 77976-842
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC 6-(Aminocarbonyl)-3-cyclohexene-1-carboxylic Acid
Catalog # 77976-842
Supplier:
LGC Standards
Supplier Number:
TRC-A357765-250MG
New Product
CAS Number:
Specifications
CAS No.:
2028-12-8
Description:
6-(Aminocarbonyl)-3-cyclohexene-1-carboxylic Acid
Size:
250 mg
Specifications
About this item
Product Details
Product Details