Dr. Ehrenstorfer Perfluorobutanoic acid-methyl ester
Catalog # 77976-067
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:356-24-1
- Description:Perfluorobutanoic acid - methyl ester
- Size:100 mg
- Shipping conditions:20 °C
- Formula:C₅H₃F₇O₂
- Formula weight:228,065 g/mol
- Storage temperature:20 °C
- Certification:ISO/IEC 17025
- Certification:ISO/IEC 17025