Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC Propofol-d17 Beta-D-Glucuronide
TRC Propofol-d17 Beta-D-Glucuronide
TRC Propofol-d17 Beta-D-Glucuronide
Catalog # 77992-239
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Propofol-d17 Beta-D-Glucuronide
Catalog # 77992-239
Supplier:
LGC Standards
Supplier Number:
TRC-P829782-5MG
New Product
CAS Number:
Specifications
Description:
Propofol-d17 Beta-D-Glucuronide
Size:
5 mg
Specifications
About this item
Product Details
Product Details