Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 1-(4-Amino-3,5-dichlorophenyl)-2-ethanone
TRC 1-(4-Amino-3,5-dichlorophenyl)-2-ethanone
TRC 1-(4-Amino-3,5-dichlorophenyl)-2-ethanone
77977-157
:
LGC Standards
New Product
:
TRC 1-(4-Amino-3,5-dichlorophenyl)-2-ethanone
77977-157
:
LGC Standards
:
TRC-A604690-500MG
New Product
:
CAS No.:
37148-48-4
Description:
1-(4-Amino-3,5-dichlorophenyl)-2-ethanone
Size:
500 mg