Paragon Scientific Sulfur @ 4% (40000 ppm) in synthetic diesel
77975-749
New Product
:
- Matrix:Synthetic Diesel
- CAS No.:7704-34-9
- Description:Sulfur @ 4% (40000 ppm) in synthetic diesel
- Size:100 ml
- Certification:ISO 17034
- Storage conditions:RT
- Shipping conditions:RT
- Molecular weight:32.065