Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC Polymyxin B2 Formate Salt
TRC Polymyxin B2 Formate Salt
TRC Polymyxin B2 Formate Salt
Catalog # 77991-982
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Polymyxin B2 Formate Salt
Catalog # 77991-982
Supplier:
LGC Standards
Supplier Number:
TRC-P689396-2.5MG
New Product
CAS Number:
Specifications
Description:
Polymyxin B2 Formate Salt
Size:
2.5 mg
Specifications
About this item
Product Details
Product Details