Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC N,N'-Bis[4-chloro-3-trifluoromethylphenyl]urea
TRC N,N'-Bis[4-chloro-3-trifluoromethylphenyl]urea
TRC N,N'-Bis[4-chloro-3-trifluoromethylphenyl]urea
Catalog # 77978-581
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC N,N'-Bis[4-chloro-3-trifluoromethylphenyl]urea
Catalog # 77978-581
Supplier:
LGC Standards
Supplier Number:
TRC-B414135-100MG
New Product
CAS Number:
Specifications
CAS No.:
370-50-3
Description:
N,N'-Bis[4-chloro-3-trifluoromethylphenyl]urea
Size:
100 mg
Specifications
About this item
Product Details
Product Details