Dr. Ehrenstorfer Microplastics Polypropylene PP nominal 100 μm) (Dv50 = 180 μm)
Catalog # 77976-078
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:9003-07-0
- Description:Microplastics polypropylene PP nominal 100 μm (Dv50 = 180 μm; Dn50 = 3 μm)
- Size:50 mg
- Shipping conditions:20 °C
- Formula:n(C₃H₆)
- Formula weight:42,08 g/mol
- Storage temperature:20 °C
- Certification:ISO/IEC 17025