Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC Vincristine-d3 Sulfate
TRC Vincristine-d3 Sulfate
TRC Vincristine-d3 Sulfate
77995-260
:
LGC Standards
New Product
:
TRC Vincristine-d3 Sulfate
77995-260
:
LGC Standards
:
TRC-V314253-1MG
New Product
:
CAS No.:
1246817-10-6
Description:
Vincristine-d3 Sulfate
Size:
1 mg