Order Entry
Canada
ContactUsLinkComponent
Paragon Scientific ASTM D7220, Sulfur set B in odorless kerosene: S @ 0.005, 0.01, 0.025, 0.05, 0.075, 0.1%
Paragon Scientific ASTM D7220, Sulfur set B in odorless kerosene: S @ 0.005, 0.01, 0.025, 0.05, 0.075, 0.1%
  77975-625
New Product
 :  
Paragon Scientific ASTM D7220, Sulfur set B in odorless kerosene: S @ 0.005, 0.01, 0.025, 0.05, 0.075, 0.1%
  77975-625
 :  ALK-D7220-KRSUB
New Product
 :  

 

  • CAS No.:
    7704-34-9
  • Storage conditions:
    RT
  • Shipping conditions:
    RT
  • Molecular weight:
    32.065