Order Entry
Canada
ContactUsLinkComponent
Paragon Scientific 2 µS/cm @ 25 °C, Conductivity (NIST Traceable, ACS)
Paragon Scientific 2 µS/cm @ 25 °C, Conductivity (NIST Traceable, ACS)
Catalog # 77975-545
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Paragon Scientific 2 µS/cm @ 25 °C, Conductivity (NIST Traceable, ACS)
Catalog # 77975-545
Supplier:  LGC Standards
Supplier Number:  ALK-COND0002-25-125ML
New Product
CAS Number:  

Specifications

  • Description:
    Conductivity standard CRM
  • Size:
    125 ml
  • Reference Temperature:
    25 °C
  • Certification:
    ISO 17034
  • Conductivity:
    2 µS/cm

Specifications

About this item