Order Entry
Canada
ContactUsLinkComponent
CHEMets® molybdate Visual Test Kit 0 to 7 ppm
CHEMets® molybdate Visual Test Kit 0 to 7 ppm
Catalog # 77674-816
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
CHEMets® molybdate Visual Test Kit 0 to 7 ppm
Catalog # 77674-816
Supplier:  CHEMetrics
Supplier Number:  K-6701
New Product
CAS Number:  

Specifications

  • Chemical method:
    Catechol
  • Application:
    Industrial water treatment;Power generation;Cooling water systems

Specifications

About this item

  • Range: 0 to 7 ppm as Mo MDL: 0.5 ppm Method: Catechol
Delivery: Kit K-6701 comes in a plastic case and contains everything needed to perform 30 tests:refill, comparator, 25 ml sample cup, and instructions.