Paragon Scientific IP 336, Sulfur Set E in synthetic diesel: S @ blank, 0.03, 0.05, 0.07, 0.1, 0.3, 0.5%
Catalog # 77975-799
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Synthetic Diesel
- CAS No.:7704-34-9
- Description:IP 336, Sulfur Set E in synthetic diesel: S @ blank, 0.03, 0.05, 0.07, 0.1, 0.3, 0.5%
- Size:7×100ml
- Certification:ISO 17034
- Storage conditions:RT
- Shipping conditions:RT
- Molecular weight:32.065