TRC Ceftriaxone
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:73384-59-5
Specifications
Frequently Bought Together
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Water, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
Chlorine Test Papers, Lamotte Chemical Products
Supplier: LaMotte
Oakton® Conductivity and TDS Standards, Cole Parmer
Supplier: Antyila Scientific
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Acetone ≥99.5% (by GC, corrected for water content), GR ACS, Supelco®
Supplier: MilliporeSigma
Leakbuster™ Specimen Containers
Supplier: Starplex
Triethylamine ≥99.5% for HPLC, Supelco®
Supplier: MilliporeSigma
VWR® PurFlock Ultra 6" Sterile Revised Ultrafine Flock Swab with Polystyrene Handle, 80 mm Breakpoint
Supplier: VWR International
BD GasPak™ EZ Container Systems, BD Diagnostics
Supplier: BD
Water for HPLC, Supelco®
Supplier: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Xylene
Supplier: Ward's Science
Tetrahydrofuran ≥99.5% (by GC, corrected for water content) stabilised, BAKER ANALYZED® A.C.S. Reagent ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC