TRC 3-Desmethoxy-3-bromo Trimethoprim
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:16285-82-8
Specifications
Frequently Bought Together
Starswabs II® Specimen Collection and Transport Systems
Supplier: Starplex
Calcium chloride dihydrate 99.0-107.0% USP, Ph. Eur., BP, JP endotoxin tested
Supplier: Avantor Performance Materials
D-(+)-Sucrose, Ultrapure DNase-, RNase-free
Supplier: VWR
D-(+)-Maltose monohydrate, Reagent Grade
Supplier: VWR
VACUETTE® Secondary Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
Water ACS, Reagent Grade ASTM Type I, ASTM Type II Packaged in plastic containers
Supplier: Ricca Chemical
COD Digestion vials, VWR Chemicals BDH®
Supplier: VWR International
SYRINGE STERILE WITHOUT NEEDLE 10ML
Supplier: AMX INDUSTRIAL TRADE MX
LabKlenz™ 100 Alkaline Detergents
Supplier: Steris
Tissue Marking Dyes, Azer Scientific
Supplier: Azer Scientific
Acetic acid glacial 99.5-100.5% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
PETROLEUM BENZINE 1L
Supplier: MilliporeSigma
1,5-Diaminopentane 98%
Supplier: Thermo Scientific Chemicals
Gastight® 1800 Series Syringes, Hamilton
Supplier: Hamilton
Sodium acetate, anhydrous ≥99.0% (by perchloric acid titration), EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma