Dr. Ehrenstorfer Perfluoro-4-ethylcyclohexylsulfonic acid potassium 50 µg/ml in Methanol:Water
Catalog # 77976-043
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol:Water
- CAS No.:335-24-0
- Description:Perfluoro-4-ethylcyclohexylsulfonic acid potassium 50 µg/ml in Methanol:Water
- Size:1 ml
- Shipping conditions:20 °C
- Formula:C₈F₁₅O₃S·K
- Formula weight:500,223 g/mol
- Storage temperature:−18 °C
- Certification:ISO 17034