TRC rac 1-Palmitoyl-3-chloropropanediol
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:30557-04-1
Specifications
Frequently Bought Together
VWR® Multipurpose Containers, HDPE
Supplier: VWR International
MQuant® pH Test Strips, Supelco®,MilliporeSigma
Supplier: MilliporeSigma
Medicine Cups, DUKAL™ Corporation
Supplier: Dukal
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Peroxide Test, MQuant®, MilliporeSigma
Supplier: MilliporeSigma
Ammonium Hydroxide
Supplier: Ward's Science
Difco™ Agar Soldifying Agent, BD Diagnostics
Supplier: BD
Dibutyryl-cAMP sodium salt ≥97.0% (by HPLC)
Supplier: TCI America
Acetic acid glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Human Recombinant CNTF (from <i>E. coli</i>)
Supplier: Peprotech
Accessories for E.Z.N.A.® Blood DNA Extraction Kits, Omega Bio-Tek
Supplier: Omega Bio-Tek
Hazmat Boot Covers, Onguard
Supplier: Onguard Industries
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane ≥99%
Supplier: Thermo Scientific Chemicals
Polyethylene glycol 4,000
Supplier: Thermo Scientific Chemicals