Order Entry
Canada
ContactUsLinkComponent
 
Dr. Ehrenstorfer 1H,1H,2H,2H-Perfluorooctanephosphonic acid
  77976-063
New Product
 :  
undefined
Dr. Ehrenstorfer 1H,1H,2H,2H-Perfluorooctanephosphonic acid
  77976-063
 :  DRE-C15987101
New Product
 :  

 

  • CAS No.:
    252237-40-4
  • Description:
    1H, 1H, 2H, 2H - Perfluorooctanephosphonic acid
  • Size:
    50 mg
  • Shipping conditions:
    20 °C
  • Formula:
    C₈H₆F₁₃O₃P
  • Formula weight:
    428,084 g/mol
  • Storage temperature:
    20 °C
  • Certification:
    ISO/IEC 17025