Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 3-(1,2,4-Triazol-1-yl)-L-alanine
TRC 3-(1,2,4-Triazol-1-yl)-L-alanine
TRC 3-(1,2,4-Triazol-1-yl)-L-alanine
77994-599
:
LGC Standards
New Product
:
TRC 3-(1,2,4-Triazol-1-yl)-L-alanine
77994-599
:
LGC Standards
:
TRC-T767515-10MG
New Product
:
CAS No.:
4819-36-7
Description:
3-(1,2,4-Triazol-1-yl)-L-alanine
Size:
10 mg